missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgG (H+L) Secondary Antibody, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen PA128787
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
PA1-28787 detects Rat IgG from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifica
Rat IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
IgG fraction | |
RUO | |
Rat | |
Liquid |
Dot Blot, ELISA, Immunohistochemistry, Immunomicroscopy, Western Blot | |
1 mg/mL | |
0.05M tris HCl with 0.15M NaCl, 0.1mM ZnCl2, 10mg/mL BSA, 1mM MgCl2, 50% glycerol and 0.01% sodium azide; pH 8.0 | |
Full length native Rat IgG (purified). | |
100 μg | |
Secondary | |
4° C, do not freeze | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto