missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG control Rabbit, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL594-30000-100UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Rabbit IgG control Polyclonal antibody apecifically detetects Rabbit IgG control antigen in Samples. It is validated for Flow Cytometry.Specifica
Rabbit IgG control | |
Polyclonal | |
CoraLite 594 | |
Rabbit | |
Primary | |
Liquid |
Flow Cytometry | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
100 μL | |
Store at -20°C. Avoid exposure to light. | |
IgG |