missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RadEye SPRD and SPRD-ER Personal Radiation Detector

Experience extremely high radiation dose rate measurement and accuracy with the Thermo Scientific™ RadEye™ SPRD-ER Personal Radiation Detector.

Marca:  Thermo Scientific™ 4250825

 vedi altre versioni di questo prodotto

Codice prodotto. 16883836

  • 5975.00€ / Pezzo
Spedizione stimata: 12-02-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso

Descrizione

Descrizione

Respond quickly and decisively to real radiation threats with the new standard in detection and interdiction of illicit radiation. The Thermo Scientific™ RadEye™ SPRD and SPRD-ER Personal Radiation Detectors are handheld radiation monitors that offer a unique combination of detector sensitivity and intelligent alarming to help ensure detection in the most challenging applications while eliminating nuisance alarms. Additionally, the RadEye SPRD and SPRD-ER radiation detectors identify and categorizes specific types of radiation or radioactive material, and can be configured to support your missions or user skills.

By providing significant detector sensitivity for search and find, combined with accurate high dose rate radiation measurement, the RadEye SPRD-ER is a suitable instrument for all users and may be the only radiation detector you need.

Ensure safety - extremely high dose rate measurement and accuracy

  • Extremely high dose rate measurement capabilities of ≤1000 R/h (<10 Sv/h) supports users with one unit for search, find, radiation identification and response missions.
  • Unprecedented dose rate accuracy across the full spectrum of 20keV – 3 MeV
  • New highly sensitive alarm for artificial gamma (S-Alarm) supplies extra sensitivity in the range of 90 - 450 keV for effective detection of SNM materials
  • Sourceless detector gain stabilization improves radiation identification selectivity and accuracy across the spectrum in varying environmental conditions.

Stay on mission - detect radiation at greater distances

  • Advanced NBR identifies artificial radiation while minimizing false alarms

Respond swiftly - faster discrimination and adjudication of potential threats

  • (SNM, Industrial) and non-threat sources (medical or natural)
  • Neutron indication via proven prompt gamma analysis

Effortless deployment

  • Simplified ID capability for novice to advanced users
  • Multiple modes of operation, easily configured to best align with radiation detection operating procedures

 

Specifica

Specifica

(662 keV)
RadEye SPRD-ER Personal Radiation Detector
Spectroscopic energy range, Low dose rate / Search / ID detector: 20 keV - 3 MeV
2 AAA alkaline or NiMH
Wireless: Bluetooth 4.0 (Option)
Low dose rate / Search / ID detector: CsI(Tl)
Low dose rate / Search / ID detector: Advanced algorithms
Low dose rate / Search / ID detector: Sourceless spectral algorithm
1.5 m on concrete (with rubber sleeve)
> 170 hr. (alkaline)
Low dose rate / Search / ID detector: Prompt gamma analysis
Low dose rate range: ANSI N42.32 PRD standard fully compliant; ANSI N42.48 SPRD standard fully compliant
4.1 x 2.6 x 1.6 in. (with rubber protective sleeve)
at 1μSv/h (100μR/h), Low dose rate / Search / ID detector: < 3 min
−20°C to +50°C
-4°F to +122°F
Low dose rate / Search / ID detector: Lutetium Testadapter (< 10 nCi Lu-176)
2
0.195 kg (including batteries and rubber sleeve)
Gamma, X-Ray plus neutrons via prompt gamma
0.429 lb. (including batteries and rubber sleeve)
Low dose rate / Search / ID detector: 10 nSv/h - 250 μSv/h (1 μR/h - 25 mR/h)
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato