Learn More
Fisherbrand™ Radiation Gamma Scientific Cabinets
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Dimensioni (L x P x A):
49 x 55 x 37cm
Dimensione della confezione:
Pezzo
Descrizione
- Two isolated chambers each with their own hinged l each chamber has removable insert 125mm x 85 x 95mm Fisherbrand
- Cleaver Scientific cabinets provide a convenient area to carry out work with beta and gamma emitting isotopes with complete all round protection
- Each cabinet's 49 x 55 x 37cm dimensions offer a large working area without impeding vision, either in a standing or seated position
- Services can also be used inside the cabinet via a rear port
- The back panel has a pipette holder to keep the work surface clear
- The unit can store radioactive materials when the hinged doors are closed
- These cabinets are compatible with 68 x 54cm trays, CSR-TY4, CSR-
- TW4 and CSR-TO4
- Sizeable ergonomic working space, option of being fully enclosed, all round user protection
Specifica
Specifica
| Dimensioni (L x P x A) | 49 x 55 x 37cm |
| Product Type | Radiaton Gamma Scientific Cabinets |