missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA552282
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: HAMNNLNGTE LEGSCLEVTL AKPVDKEQYS RYQKAARGGG AAEAAQQPSY VYSCDPYTLA YYGYPYNALI GPNRDYFVKA GSIRGRGRGA AGNRAPGPRG SYLGGYSAGR GIYSRYHEGK GKQQEKGYEL VPNLEIPTVN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The function of this protein remains unknown.Specifica
RBM47 | |
Polyclonal | |
Unconjugated | |
RBM47 | |
9530077J19Rik; BC61; hypothetical protein; hypothetical RNA binding protein RGD1359713; NET18; RBM47; RGD1359713; RNA binding motif protein 47; RNA-binding motif protein 47; RNA-binding protein 47; si:ch211-241e15.2; wu:fj94a03 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
305340, 54502 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0AV96, Q66H68 | |
RBM47 | |
Recombinant protein corresponding to Human RBM47 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only