Learn More
DiscoverX Ready-to-Assay™ ETA Endothelin Receptor Frozen Cells
The Ready-to-Assay™ ETA Endothelin Receptor Frozen Cells measure EDNRA (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.
Marca: DiscoverX HTS070RTA
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The Ready-to-Assay™ ETA Endothelin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. endothelin) based activation of EDNRA GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress EDNRA alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of EDNRA , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.Specifica
ETA | |
endothelin | |
EDNRA |
Frozen Cells | |
2 Vials of Cells |