missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis-inducing factor 1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis-inducing factor 1, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5633-ec-500ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_001124318.2 | |
Apoptosis-inducing factor 1, mitochondrial Protein | |
Research Use Only | |
AIFM1 | |
Recombinant Protein | |
E. coli | |
GLTPEQKQKK AALSASEGEE VPQDKAPSHV PFLLIGGGTA AFAAARSIRA RDPGARVLIV SEDPELPYMR PPLSKELWFS DDPNVTKTLR FKQWNGKERS IYFQPPSFYV SAQDLPHIEN GGVAVLTGKK VVQLDVRDNM VKLNDGSQIT YEKCLIATGG TPRSLSAIDR AGAEVKSRTT LFRKIGDFRS LEKISREVKS ITIIGGGFLG SELACALGRK ARALGTEVIQ LFPEKGNMGK ILPEYLSNWT MEKVRREGVK VMPNAIVQSV GVSSGKLLIK LKDGRKVETD HIVAAVGLEP NVELAKTGGL EIDSDFGGFR VNAELQARSN IWVAGDAACF YDIKLGRRRV EHHDHAVVSG RLAGENMTGA AKPYWHQSMF WSDLGPDVGY EAIGLVDSSL PTVGVFAKAT AQDNPKSATE QSGTGIRSES ETESEASEIT IPPSTPAVPQ APVQGEDYGK GVIFYLRDKV VVGIVLWNIF NRMPIARKII KDGEQHEDLN EVAKLFNIHE | |
Greater than 90% as determined by SDS-PAGE. |
9131 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |