missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AZGP1 Protein
A cDNA sequence encoding the AZGP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11114-1ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
563 | |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MGSSHHHHHH SSGLVPRGSH MGSQENQDGR YSLTYIYTGL SKHVEDVPAF QALGSLNDLQ FFRYNSKDRK SQPMGLWRQV EGMEDWKQDS QLQKAREDIF METLKDIVEY YNDSNGSHVL QGRFGCEIEN NRSSGAFWKY YYDGKDYIEF NKEIPAWVPF DPAAQITKQK WEAEPVYVQR AKAYLEEECP ATLRKYLKYS KNILDRQDPP SVVVTSHQAP GEKKKLKCLA YDFYPGKIDV HWTRAGEVQE PELRGDVLHN GNGTYQSWVV VAVPPQDTAP YSCHVQHSSL AQPLVVPWEA S | |
Greater than 85.0% as determined by SDS-PAGE. |
AZGP1 Protein | |
Research Use Only | |
AZGP1 | |
Human | |
Untagged | |
AZGP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |