missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FTSJ1 Protein
A DNA sequence encoding the Homo sapiens (Human) FTSJ1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6058-ec-200ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_001269086.1 | |
FTSJ1 Protein | |
Research Use Only | |
FTSJ1 | |
Recombinant Protein | |
E. coli | |
MGRTSKDKRD VYYRLAKENG WRARSAFKLL QLDKEFQLFQ GVTRAVDLCA APGSWSQVLS QKIGGQGSGH VVAVDLQAMA PLPGVVQIQG DITQLSTAKE IIQHFKGCPA DLVVCDGAPD VTGLHDVDEY MQAQLLLAAL NIATHVLKPG GCFVAKIFRG RDVTLLYSQL QVFFSSVLCA KPRSSRNSSI EAFAVCQGYD PPEGFIPDLS KPLLDHSYDP DFNQLDGPTR IIVPFVTCGD LSSYDSDRSY PLDLEGGSEY KYTPPTQPPI SPPYQEACTL KRKGQLAKEI RPQDCPISRV DTFPQPLAAP QCHTLLAPEM EDNEMSCSP | |
Greater than 90% as determined by SDS-PAGE. |
24140 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |