Learn More
enQuireBio™ Recombinant Human HDAC3 / Histone deacetylase 3 Protein
Codice prodotto. 16071190
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
Codice prodotto. 16071190
Marca: enQuireBio™ QP6152ec100ug
A DNA sequence encoding the Homo sapiens (Human) HDAC3 / Histone deacetylase 3, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso NP_003874.2
ID gene (immissione) 8841
Nome HDAC3 / Histone deacetylase 3 Protein
Quantità 100 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene HDAC3
Lunghezza della proteina Full Length
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His-SUMO
Sequenza MAKTVAYFYD PDVGNFHYGA GHPMKPHRLA LTHSLVLHYG LYKKMIVFKP YQASQHDMCR FHSEDYIDFL QRVSPTNMQG FTKSLNAFNV GDDCPVFPGL FEFCSRYTGA SLQGATQLNN KICDIAINWA GGLHHAKKFE ASGFCYVNDI VIGILELLKY HPRVLYIDID IHHGDGVQEA FYLTDRVMTV SFHKYGNYFF PGTGDMYEVG AESGRYYCLN VPLRDGIDDQ SYKHLFQPVI NQVVDFYQPT CIVLQCGADS LGCDRLGCFN LSIRGHGECV EYVKSFNIPL LVLGGGGYTV RNVARCWTYE TSLLVEEAIS EELPYSEYFE YFAPDFTLHP DVSTRIENQN SRQYLDQIRQ TIFENLKMLN HAPSVQIHDV PADLLTYDRT DEADAEERGP EENYSRPEAP NEFYDGDHDN DKESDVEI
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.