missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Polypyrimidine tract-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Polypyrimidine tract-binding protein 1, was expressed in the hosts and tags indicated.
710.00€ - 1985.00€
Specifica
Numero di accesso | NP_002810.1 |
---|---|
ID gene (immissione) | 5725 |
Nome | Polypyrimidine tract-binding protein 1 |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not determined. |
Specifica
NP_002810.1 | |
Polypyrimidine tract-binding protein 1 | |
Not determined. | |
Full length of Isoform 3 | |
Human | |
Flag | |
Tris-based buffer, 50% glycerol |
5725 | |
Research Use Only | |
PTBP1 | |
Recombinant Protein | |
Mammal | |
MDGIVPDIAV GTKRGSDELF STCVTNGPFI MSSNSASAAN GNDSKKFKGD SRSAGVPSRV IHIRKLPIDV TEGEVISLGL PFGKVTNLLM LKGKNQAFIE MNTEEAANTM VNYYTSVTPV LRGQPIYIQF SNHKELKTDS SPNQARAQAA LQAVNSVQSG NLALAASAAA VDAGMAMAGQ SPVLRIIVEN LFYPVTLDVL HQIFSKFGTV LKIITFTKNN QFQALLQYAD PVSAQHAKLS LDGQNIYNAC CTLRIDFSKL TSLNVKYNND KSRDYTRPDL PSGDSQPSLD QTMAAAFGAP GIISASPYAG AGFPPTFAIP QAAGLSVPNV HGALAPLAIP SAAAAAAAAG RIAIPGLAGA GNSVLLVSNL NPERVTPQSL FILFGVYGDV QRVKILFNKK ENALVQMADG NQAQLAMSHL NGHKLHGKPI RITLSKHQNV QLPREGQEDQ GLTKDYGNSP LHRFKKPGSK NFQNIFPPSA TLHLSNIPPS VSEEDLKVLF SSNGGVVKGF KFFQKDRKMA LIQMGSVEEA VQALIDLHNH DLGENHHLRV SFSKSTI | |
Greater than 90% as determined by SDS-PAGE. |