missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-8A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-8A, was expressed in the hosts and tags indicated.
160.00€ - 1855.00€
Specifica
Numero di accesso | NP_005361.2 |
---|---|
ID gene (immissione) | 4218 |
Nome | Ras-related protein Rab-8A Protein |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not determined. |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16054270
|
enQuireBio™
QP6575-ec-10ug |
10 μg | |||||||
16094270
|
enQuireBio™
QP6575-ec-50ug |
50 μg | |||||||
16044270
|
enQuireBio™
QP6575-ec-100ug |
100 μg | |||||||
16074270
|
enQuireBio™
QP6575-ec-200ug |
200 μg | |||||||
16084270
|
enQuireBio™
QP6575-ec-500ug |
500 μg | |||||||
16064270
|
enQuireBio™
QP6575-ec-1mg |
1 mg | |||||||
Specifica
NP_005361.2 | |
Ras-related protein Rab-8A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4218 | |
Research Use Only | |
RAB8A | |
Recombinant Protein | |
E. coli | |
KTYDYLFKLL LIGDSGVGKT CVLFRFSEDA FNSTFISTIG IDFKIRTIEL DGKRIKLQIW DTAGQERFRT ITTAYYRGAM GIMLVYDITN EKSFDNIRNW IRNIEEHASA DVEKMILGNK CDVNDKRQVS KERGEKLALD YGIKFMETSA KANINVENAF FTLARDIKAK MDKKLEGNSP QGSNQGVKIT P | |
Greater than 90% as determined by SDS-PAGE. |