missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trichoplein keratin filament-binding Protein
A DNA sequence encoding the Homo sapiens (Human) Trichoplein keratin filament-binding protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8130-ec-200ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_001137324.1 | |
Trichoplein keratin filament-binding Protein | |
Research Use Only | |
TCHP | |
Recombinant Protein | |
E. coli | |
MALPTLPSYW CSQQRLNQQL ARQREQEARL RQQWEQNSRY FRMSDICSSK QAEWSSKTSY QRSMHAYQRE KMKEEKRRSL EARREKLRQL MQEEQDLLAR ELEELRLSMN LQERRIREQH GKLKSAKEEQ RKLIAEQLLY EHWKKNNPKL REMELDLHQK HVVNSWEMQK EEKKQQEATA EQENKRYENE YERARREALE RMKAEEERRQ LEDKLQAEAL LQQMEELKLK EVEATKLKKE QENLLKQRWE LERLEEERKQ MEAFRQKAEL GRFLRHQYNA QLSRRTQQIQ EELEADRRIL QALLEKEDES QRLHLARREQ VMADVAWMKQ AIEEQLQLER AREAELQMLL REEAKEMWEK REAEWARERS ARDRLMSEVL TGRQQQIQEK IEQNRRAQEE SLKHREQLIR NLEEVRELAR REKEESEKLK SARKQELEAQ VAERRLQAWE ADQQEEEEEE EARRVEQLSD ALLQQEAETM AEQGYRPKPY GHPKIAWN | |
Greater than 90% as determined by SDS-PAGE. |
84260 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |