missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human U6 snRNA-associated Sm-like protein LSm4 Protein
A DNA sequence encoding the Homo sapiens (Human) U6 snRNA-associated Sm-like protein LSm4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6322-ec-100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_036453.1 | |
U6 snRNA-associated Sm-like protein LSm4 Protein | |
Research Use Only | |
LSM4 | |
Recombinant Protein | |
E. coli | |
MLPLSLLKTA QNHPMLVELK NGETYNGHLV SCDNWMNINL REVICTSRDG DKFWRMPECY IRGSTIKYLR IPDEIIDMVK EEVVAKGRGR GGLQQQKQQK GRGMGGAGRG VFGGRGRGGI PGTGRGQPEK KPGRQAGKQ | |
Greater than 90% as determined by SDS-PAGE. |
25804 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |