missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Transmembrane protease serine 4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Transmembrane protease serine 4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7925-ec-50ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_663378.1 | |
Transmembrane protease serine 4 Protein | |
Research Use Only | |
Tmprss4 | |
Recombinant Protein | |
E. coli | |
KVILDKYYFI CGSPLTFIQR GQLCDGHLDC ASGEDEEHCV KDFPEKPGVA VRLSKDRSTL QVLDAATGTW ASVCFDNFTE ALAKTACRQM GYDSQPAFRA VEIRPDQNLP VAQVTGNSQE LQVQNGSRSC LSGSLVSLRC LDCGKSLKTP RVVGGVEAPV DSWPWQVSIQ YNKQHVCGGS ILDPHWILTA AHCFRKYLDV SSWKVRAGSN ILGNSPSLPV AKIFIAEPNP LYPKEKDIAL VKLQMPLTFS GSVRPICLPF SDEVLVPATP VWVIGWGFTE ENGGKMSDML LQASVQVIDS TRCNAEDAYE GEVTAEMLCA GTPQGGKDTC QGDSGGPLMY HSDKWQVVGI VSWGHGCGGP STPGVYTKVT AYLNWIYNVR KSEM | |
Greater than 90% as determined by SDS-PAGE. |
214523 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |