missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL11 / Eotaxin Protein
A DNA sequence encoding the Rat CCL11 / Eotaxin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10296-ec-250ug
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_062078.1 | |
CCL11 / Eotaxin Protein | |
Research Use Only | |
CCL11 | |
The biological activity determined by a chemotaxis bioassay using purified eosinophils is in a concentration range of 0.1-1.0 ug/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
29397 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
HPGSIPTSCC FTMTSKKIPN TLLKSYKRIT NNRCTLKAIV FKTKLGKEIC ADPKKKWVQD ATKHLDQKLQ TPKP | |
Greater than 96% as determined by SDS-PAGE and HPLC. |