missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL-1 beta / IL1B Protein

Product Code. 15936319
missing translation for 'orderingAttributeHoverText'
Quantità:
1 mg
10 μg
100 μg
2 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
2µg
500µg
This item is not returnable. View return policy

Product Code. 15936319

missing translation for 'mfr': enQuireBio™ QP55161mg

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Nome IL-1 beta / IL1B Protein
Quantità 1 mg
Status giuridico Research Use Only
Concentrazione di endotossine Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Simbolo del gene IL1B
Attività biologica Rat IL-1 beta / IL1B induces D10S cell proliferation (typical ED50 is ? 25 ng/ml).
Product Type Recombinant Protein
Cross-reattività Rat
Specie E. coli
Tag proteine Untagged
Sequenza MVPIRQLHCR LRDEQQKCLV LSDPCELKAL HLNGQNISQQ VVFSMSFVQG ETSNDKIPVA LGLKGLNLYL SCVMKDGTPT LQLESVDPKQ YPKKKMEKRF VFNKIEVKTK VEFESAQFPN WYISTSQAEH RPVFLGNSNG RDIVDFTMEP VSS
Tampone 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.