Learn More
Abnova™ RedTaq DNA Polymerase
Codice prodotto. 16042555
Click to view available options
Quantità:
1000 units
2500 units
500 units
Dimensione della confezione:
1000 unità
2500 unità
500 unità
Codice prodotto. 16042555
Marca: Abnova™ D0012.500Units
RedTaq DNA polymerase catalyzes 5'-3' synthesis of DNA.
  • The enzyme has been proved not having the 3′-5′ exonuclease activity.
  • The enzyme has proven to have high amplification yield, be stable at high temparetaure.
  • Added inert dye will not have any interference to the reaction.
  • Visual confirmation that the enzyme has been added and proper component mixing of the reaction has occurred.
  • Samples can be loaded directly onto an agarose gel for electrophoresis with no loading dye addition.
Specifica
Concentrazione 1 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantità 500 units
Da utilizzare con (applicazione) Standard PCR
Tampone di conservazione 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol