Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile D/E (Dey-Engley) Neutralizing Agar (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile D/E Neutralizing Agar (Irradiated) is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
Marca: Thermo Scientific™ R111803
143.73 EUR valido fino al 2025-01-31
Usa il codice promo "23894" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Thermo Scientific™ Remel Contact Plate Sterile D/E Neutralizing Agar (Irradiated) is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
- Contact Plates Sterile are double-bagged and a third bag is enclosed for customer use (Double + Bag)
- These products are manufactured in a clean room and validated sterile by gamma radiation
- The Contact Plates are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifica
Contact Plates, D/E-Neutralizing Agar | |
Contact - Double Bag | |
Agar | |
10/Pk. |
Not intended for IVD use.