Learn More
Thermo Scientific™ Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).
Marca:  Thermo Scientific™ R111874
Dettagli aggiuntivi : Peso : 0.70000kg
Codice prodotto. 17415539
  • 232.00€ / pezzi 20
Spedizione stimata: 06-09-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready-to-use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifica
Specifica
Settling Plate Sterile Tryptic Soy Agar
15mm x 150mm Plate - Double + Bag
Agar
20/Pk.
Offerte speciali
Not intended for IVD use.