Learn More
Labcon Reusable Racks For Centrifuge Tubes
Codice prodotto. p-7236559 Sfoglia Tutto Labcon Prodotti
Click to view available options
Colore:
Assorted
Blue
Green
Natural
Orange
Purple
Red
White
Yellow
Da utilizzare con (apparecchiatura):
15 mL Centrifuge, 5 mL Snap Cap Tubes
50 mL Centrifuge Tubes
Dimensione della confezione:
pezzi 5
Codice prodotto. 17494342
Marca: Labcon 27152200009
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Plastic centrifuge tube storage racks eliminate wasteful styrofoam racks. They are durable, reusable, interlockable, stackable, freezable and recyclable. Made from 100% pre-consumer, high quality recycled polypropylene from our in-house scrap recycling program.
Designed to hold Labcon™ tubes specifically, but may fit other brands of centrifuge tubes.
  • Holds (25) 15 or 50 mL Conical Centrifuge Tubes
  • Includes Numbered Wells
  • Racks Lock Together To Form Work Stations
  • Replaces Disposable Polystyrene Foam Racks
  • Made of Polypropylene
  • Freezable to -80°C
  • Autoclavable
Specifica
Include Racks
Product Type Test Tube Rack
Matrice 25
Autoclavabile Autoclavable
Colore Natural
Capacità 25 Tubes
Materiale Polypropylene
Sovrapponibile
Sterilità Non-sterile
Da utilizzare con (apparecchiatura) 15 mL Centrifuge, 5 mL Snap Cap Tubes
Intervallo di temperatura (sistema metrico) Freezable (-80°C)
Dimensioni (L x P) 14.88 x 11.7 cm
Altezza (sistema metrico) 3.81 cm