Learn More
Thermo Scientific™ RheBuild™ Kits
Codice prodotto. 10149464 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7010/7000
Rheodyne Injectors 7125/7126
Rheodyne Injectors 7725/7725i/7726
Rheodyne Injectors 8125/8126
Rheodyne Injectors 9125/9126
Dimensione della confezione:
Pezzo
Codice prodotto. 10149464
Marca: Thermo Scientific™ 3725999
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Everything you need to maintain your Thermo Scientific™ Rheodyne™ valves and injectors in your HPLC system.
Use Thermo Scientific™ RheBuild Kits to maintain your Thermo Scientific™ Rheodyne™ valves and injectors for high-performance liquid chromatography (HPLC). A single kit includes everything you need.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Maintenance Kit
Descrizione RheBuild Kit
Da utilizzare con (apparecchiatura) Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.