Learn More
Epredia™ Richard-Allan Scientific™ Elastic Stain, Van Gieson Stain Solution
Sfoglia Tutto Epredia Prodotti
Click to view available options
Tipo:
Alcoholic Hematoxylin Solution
Sodium Thiosulfate Solution
Van Gieson Stain
Weigert's Iodine Solution
Dimensione della confezione:
125mL
250mL
500mL
Descrizione
- Based on the Verhoeff Elastic technique
- Also used for identifying the breaks, splitting and reduplication of elastic lamellae that occur in vascular diseases
- Elastic fibers are stained blue-black to-black with red collagen and a yellow background
Specifica
Specifica
| Tipo | Sodium Thiosulfate Solution |