Learn More
Brady™ Etichette Rigid DuraSleeve per M710 M610 M611
Inserti rigidi DuraSleeve per M611, M610 e M710
Marca: Brady™ 312076
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Brady™ DuraSleeve™ printable inserts create a durable legend
- Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
- Legend can be changed before or after termination
Specifica
M6-23x4.4-7696-WT | |
White | |
Tape | |
Wire and Cable Markers | |
Wire and Cable Labeling | |
1 Label | |
-40°F to +158°F | |
0.0098 in. | |
4.4 x 23 mm | |
0.91 in. |
Non-adhesive | |
Gloss | |
Vinyl | |
Non-sterile | |
M610, M710, M611, BMP61, BMP71 | |
250 Labels | |
-40°C to +70°C | |
0.249 mm | |
0.17 in. | |
23 mm |