Learn More
Tocris Bioscience™ Ro 5-3335
Core binding factor inhibitor; attenuates hematopoiesis
Marca: Tocris Bioscience™ 4694/10
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Ro 5-3335 is a core binding factor (CBF) inhibitor; preferentially kills human leukemia cell lines with CBF fusion proteins (IC50 = 1.1 μM). Represses RUNX1/CBFβ-dependent transactivation in reporter assays and inhibits transcriptional regulation by RUNX1 and CBFβ. Reduces leukemia burden in a mouse model. Attenuates RUNX1-dependent hematopoiesis in zebrafish embryos. Also a Tat antagonist; inhibits HIV-1 replication in vitro.Specifica
>98% | |
10 mg | |
259.69 |
CBFβ-Runx1 inhibitor II | |
Ro 5-3335 | |
Ro 5-3335 |