Learn More
SI Analytics™ Rod Stirrer
SI Analytics™ Rod Stirrer for use with titration heads TZ 1463 and TZ 1467.
Brand: SI Analytics™ 285215175
Additional Details : Peso : 0.15000kg
Description
- Short model
- With NS 14.5
- For titration heads TZ 1463 and TZ 1467
Specifications
Short Model | |
Magnetic Stirrer | |
NS 14.5 |