missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rofecoxib
Click to view available options
Quantità:
250 mg
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Materiale o nome chimico | Rofecoxib |
CAS | 162011-90-7 |
Quantità | 250 mg |
Solubilità | soluble in chloroform, DMSO |
Formula molecolare | C17 H14 O4 S |
Peso formulazione | 314.36 |
Forma fisica | Crystalline Powder |
Product Type | Reagent |
Purezza | ≥97.5% (GC) |
InChI Key | RZJQGNCSTQAWON-UHFFFAOYSA-N |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'