Learn More
Ismatec™ Rotary Piston Pump Head
Click to view available options
Portata:
0.002 to 100 mL/min.
0.45 to 3400 mL/min
0002 to 100 mL/min
Dimensione della confezione:
Pezzo
Descrizione
- Coated aluminum pump-head
- Can be dismantled for cleaning
- Self-centering tube-track thanks to concave tube-bed and convex rollers, which lengthens the tube-life
- 3 stainless steel rollers (less pulsation but lower max. flow rate than with 2 rollers)
- For tubing with 1.6 mm wall thickness
- 1.5 bar differential pressure
Specifica
Specifica
| Portata | 0.002 to 100 mL/min. |
| Pressione massima | 1.0 bar |