missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labnet™ Rotisserie for Mini LabRoller™ Rotator
Rotisserie for 10 x 15 mL (16 to 17mm) tubes, 12 x 5 mL (12 to 13mm) tubes, or two microplates.
Marca: Labnet™ H5500-15
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Simple, yet versatile, the end over end tube rotator Mini LabRoller™ Rotator is designed for mixing samples in both a horizontal and vertical plane.
- Compact design
- Wide selection of motions
- Noiseless operation
- Cold room compatible
- The unit accepts a broad assortment of tube sizes as well as micro plates. Safe for cold room or incubator use, this mixer is ideal for a wide range of applications in areas such as biochemistry, molecular biology, clinical testing and histochemistry.
- H5500-15 Rotisserie accommodates ten conical 15 mL or other tubes with a 15 to 17 mm diameter as well as twelve tubes with a 12 to 13 mm diameter.
- Wide range of applications
Specifica
24 rpm | |
21.3 x 10.2 x 12.6 cm (8.4 x 4 x 5 in.) | |
Mini LabRoller™ Rotator |
10 x 15 mL Tubes (15 to 17 mm), 15 x 15 mL Tubes (12 to 13 mm) | |
Rotating Microcentrifuge Tubes | |
Up to 85% for Temperatures 4°C to 55°C |