missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Round Bottom Flasks
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 07035071
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Acc. to DIN ISO 24450 and in addition to standard
- With beaded rim
- Material: DURAN™
Specifica
65 mm | |
1 L | |
Duran Glass | |
200 mm |