missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ Round Selective Sorting Dustbin
Round Selective Sorting Dustbin is great as a highly durable recycling bin. Reinforced bottom with anti-wear skirt.
Marca: Gilac™ G613233-1
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Round dustbin great as a recycling bin.
- Highly durable.
- Reinforced bottom with anti-wear skirt.
- Reinforced handles to prevent distortion during carrying.
- Optional, choice of two lids.
- External Diameter includes handles.
- Width corresponds to diameter without handles.
Specifica
50 L | |
24 in. | |
Red | |
Round | |
18.3 in. | |
Dust Bin |
610 mm | |
13.2 gal. | |
465 mm | |
Not Autoclavable | |
HDPE |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur