Learn More
Eppendorf™ SafeCode Plates
Plates with SafeCode barcoding enable safe and reliable sample identification at the bench. Different types of plates available.
Marca: Eppendorf™ 0030113560
Dettagli aggiuntivi : Peso : 0.79200kg
Descrizione
- 3-level coding for safe sample and vessel identification
- Codes on three sides of plates for flexible reading
- ECC 200 error correction function enables safe code reading with up to 30% damaged codes
- Additional, ID-specific information on plates available via Eppendorf™ DataPort for documentation purposes
- Direct import into eLabInventory and eLabJournal software for convenient handling
- Import to other database systems possible
- Manufactured from high-quality polypropylene for highest sample purity
Specifica
Includes Barcode | |
8 x 12 | |
Clear | |
Skirted | |
250 μL | |
Sterile | |
Conical Bottom (V-well) | |
PCR Ready |
SBS Standard | |
Eppendorf | |
PCR Plate | |
Polypropylene | |
96 | |
Non-treated | |
25 Plates |