missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Saline 0.45% Solution Tubes
Reduce sample preparation time for automated identification and susceptibility systems using convenient, pre-filled and ready-to-use 0.45% saline tubes.
Marca: Thermo Scientific™ R064462
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Save time when preparing your samples for automated identification and susceptibility systems using Thermo Scientific™ Saline 0.45% Solution Tubes.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes.- Easily adjust inoculum density with a clear tube
- Enhance sample security while promoting safe handling with a reliable screw cap on each tube
- Achieve versatility and flexibility with fill volumes in 1.8mL and 3.0mL
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Clear liquid colorless | |
Saline 0.45% Solution Tubes | |
United States | |
1.8 mL | |
100/Pk. |
2 to 25°C | |
Tube | |
Buffer Solution | |
Glass tube with screw cap |