Learn More
Eppendorf™ Secondary UV-VIS-Filter
Programmed methods: ssDNA, dsDNA, RNA, oligo, protein, OD600 and Lowry, Bradford and BCA optional background correction at 320nm for UV methods
Marca:  Eppendorf™ 6131928007
Dettagli aggiuntivi : Peso : 0.37800kg
Codice prodotto. 10104323
  • 1705.00€ / Pezzo
Spedizione stimata: 14-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Programmed methods: ssDNA, dsDNA, RNA, oligo, protein, OD600 and Lowry, Bradford and BCA optional background correction at 320nm for UV methods. Automatic calculation of sample dilution. Simple conversion using the 'Conversion' key. Up to 10 standards for colorimetric processes. Xenon flash lamp with a service life of 10 years.
  • Compact unit
  • Results available at a glance
  • Large, easy to read display
  • No warm up time required
  • Measurement takes only 2 seconds
  • Storage of last 100 results and all corresponding data
  • Stored sample data and calibration data can be recalled
  • UV-vis filter set for checking photometric precision and the wavelength accuracy
Order Info
Type
Specifica
Specifica
Secondary UV-Vis Filter