Learn More
3M™ SecureFit™ SF600 Series Safety Glasses
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Antiappannamento, Antigraffio
Antigraffio
Scratch-resistant
Lens Tint:
Ambra
Clear
Gray
Grigio
I/O Mirror
Ombra saldatura 1.7
Ombra saldatura 3.0
Trasparente
Welding shade 5.0
Tipo di lente:
Polarizzati
Descrizione
- Optical class 1 lens suitable for prolonged use
- Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
- Offers protection against Ultraviolet (UV) radiation in accordance with European standards
- Weight: 27g
- 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
- Soft nose pads for comfortable fit
- Superior Scotchgard™ anti-fog coating meeting K and N on the standard clear, gray, and amber lenses
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Specifica
Lens Coating | Antigraffio |
Lens Tint | Trasparente |
Colore telaio | Grigio |
Lens Material | Policarbonato |
Materiale del telaio | Policarbonato |
UV Protection | Sì |