Learn More
SI Analytics™ Septum
SI Analytics™ Septum is an important part of the titration process and equiptment.
Marca: SI Analytics™ 285216739
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- For titration
- Accessory piece
- For Karl Fischer™ titrator
Specifica
Septa | |
Karl Fischer™ TZ 1770 and TZ 1772 Titration Vessels |