Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Codice prodotto. p-3975049
Click to view available options
Volume (sistema metrico):
20 to 500 μL
5 μL
50 μL
500 μL
Stile del punto dell'ago:
Bevel
Without Needle
Quantità:
3/Pk.
Ea.
Dimensione della confezione:
Pezzo
pezzi 3
Codice prodotto. 12927586
Marca: SGE™ 2910020
Improves laboratory workflow
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen
Applications:
  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution
Specifica
Calibro dell'ago 25 G
Diametro (sistema imperiale) interno ago 0.0047 in.
Diametro (sistema metrico) interno ago 0.12 mm
Diametro (sistema imperiale) ago esterno 0.019 in.
Diametro (sistema metrico) ago esterno 0.5 mm
Da utilizzare con (applicazione) Preparation of calibration standards, addition of internal standards
Lunghezza (sistema imperiale) ago 1.96 in.
Product Type eVol Analytical Syringe
Quantità Ea.
Volume (sistema metrico) 5 μL
Stile del punto dell'ago Bevel
Da utilizzare con (apparecchiatura) eVol Automated Syringe