Learn More
Trajan™ eVol™ MEPS™ Syringes
For use with eVol™ Digital Analytical Syringe digitally controlled positive displacement dispensing system.
Marca: Trajan™ 2910028
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Enables MEPS (micro SPE) to be semi-automated and proofed before transition to fully automated platforms
- 2011 Firmware required
- Maximum operating temperature of 350°C
Specifica
Liquid handling procedure | |
eVol™ Syringe | |
MEPS BINs |