Learn More
Trajan™ Replacement Kit for Plunger-in-Needle Syringes
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
Marca:  Trajan™ 034059
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 10656601
  • 63.70€ / Pezzo
Spedizione stimata: 10-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched. Needles and plungers must be replaced as a set
Specifica
Specifica
23 G
0.63 mm
1 μL
Syringes (1μL)
0.63 mm
Replacement Needle Kit
Cone