Learn More
Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Sfoglia Tutto Epredia Prodotti
Click to view available options
Product Type:
Formaldehyde Neutralizer Pad
Lunghezza (sistema imperiale):
10 in.
20 in.
Larghezza (sistema imperiale):
8 in.
16 in.
Dimensione della confezione:
pezzi 100
pezzi 400
pezzi 50
Descrizione
- Fiber resists tearing, even when saturated
- Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
- Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags
Specifica
Specifica
| Product Type | Formaldehyde Neutralizer Pad |
| Tipo di filtro | Formaldehyde |
| Lunghezza (sistema imperiale) | 10 in. |
| Quantità | 50/Cs. |
| Larghezza (sistema imperiale) | 8 in. |
| Larghezza (sistema metrico) | 203.2 mm |