missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ SICCO™ Big Star Desiccator Black
Big Star Desiccator Black made of black acrylic glass with 0% light transmission.
Marca: Bohlender™ V1939-07
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Aluminum frame, panels made of black acrylic glass to prevent light incidence and protect against ultraviolet radiation
- High-gloss surfaces
- Includes two shelves made of stainless steel, tray and hygrometer, usable with up to 8 shelves
- Black, absolutely lightproof panels
- For storage of light-sensitive substances
- Door with magnetic catch and circular rubber seal
- Variable height shelves
- Easy to read electronic hygrometer
- Telescopic shelves
- 0% light transmission
- Impervious to ultraviolet radiation
- Non-aging and weather-proof
Specifica
Black | |
41 gal. | |
22 in. | |
22 in. | |
23 in. | |
40 lb. |
Acrylic | |
156 L | |
560 mm | |
560 mm | |
580 mm | |
18 kg |