Learn More
Bohlender™ SICCO™ Big Star Desiccator Low
Big Star Desiccator Low made of acrylic glass and aluminum frame.
Marca:  Bohlender™ V1897-07
Codice prodotto. 17117724
  • 2275.00€ / Pezzo
Spedizione stimata: 29-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Aluminum frame with panels made of acrylic glass
  • Includes one shelf made of stainless steel, tray and hygrometer
  • Usable with up to 5 shelves, stackable
  • Reinforced aluminum frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Non-slip rubber feet
  • Easy to read electronic hygrometer
  • Variable height shelf made of stainless steel
  • Telescopic shelf
  • Desiccant tray
Specifica
Specifica
Acrylic
91 L
550 mm
330 mm
580 mm
13 kg
24 gal.
22 in.
13 in.
23 in.
29 lb.
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.