Learn More
BRAND™ SilberBrand™ ETERNA Type 2 Graduated Pipettes
AR-Glas™ graduated pipettes
Brand: BRAND™ 27010
Additional Details : Peso : 0.22500kg
Description
Specifications
To Deliver | |
B | |
Upper end cotton plug | |
Blue | |
360 mm |
DIN EN ISO 835 | |
0.1 mL | |
12 | |
5 mL |