Learn More
Thermo Scientific™ SilTite™ Replacement Ferrules , Nuts and Baseplate Seals
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal
- expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation
Specifica
Specifica
| Tipo | Replacement Ferrule |
| Descrizione | 290 MA203 |
| Da utilizzare con (apparecchiatura) | All SilTite Kits |
| Materiale | SilTite Metal |
| Include | Pk. of 10 Ferrules |
| Diameter (Metric) Inside | 0.53 mm |