Learn More
Simport™ Scientific Sample Tubes with External Threads - Graduated
With white marking area
Specifica
| Materiale chiusura | Polyethylene |
|---|---|
| Autoclavabile | Autoclavable |
| Tipo di chiusura | Screw Cap |
| Materiale | Polypropylene |
| Sterilità | Non-sterile |
Descrizione
- Made of polypropylene
- Smooth and uniform inner surface with reduced risk of contamination
- Can be autoclaved at upto 121°C in upright position with caps loosened
- Height of tube is with cap
- Available in choice of self standing or with round bottom
- Caps sold separately
- Round bottom tubes can be can be centrifuged up to 17,000g
Specifica
| Polyethylene | |
| Screw Cap | |
| Non-sterile | |
| External thread, graduations and white writing area |
| Autoclavable | |
| Polypropylene | |
| Storage and transportation of biological materials |