missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17594-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The specific function of the protein remains unknown.Specifica
SMYD4 | |
Polyclonal | |
Unconjugated | |
SMYD4 | |
KIAA1936, SMYD4, ZMYND21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114826, 287525, 319822 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BTK5, Q8IYR2 | |
Smyd4 | |
SMYD4 Fusion Protein Ag11552 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |