Learn More
Invitrogen™ SNAP91 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA550326
Codice prodotto. 15674255
  • 520.00€ / 100µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The antibody detects endogenous levels of total SNAP-91 protein.
Clathrin coat assembly protein AP180 is a protein that in humans is encoded by the SNAP91 gene. Adaptins are components of the adapter complexes which link clathrin to receptors in coated vesicles. Clathrin-associated protein complexes are believed to interact with the cytoplasmic tails of membrane proteins, leading to their selection and concentration. Binding of AP180 to clathrin triskelia induces their assembly into 60-70 nm coats.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
SNAP91
Polyclonal
Unconjugated
Snap91
91 kDa synaptosomal-associated protein; 91kDa; Ap180; assembly protein 180 (AP180); assembly protein, 180kDa; CALM; Clathrin coat assembly protein AP180; clathrin coat-associated protein AP180; F1-20; KIAA0656; mKIAA0656; phosphoprotein F1-20; SNAP91; synaptosomal-associated protein 91; synaptosomal-associated protein 91 kDa; synaptosomal-associated protein, 91 kDa; synaptosomal-associated protein, 91kDa homolog; synaptosome associated protein 91; synaptosome associated protein 91kDa
Rabbit
Antigen affinity chromatography
RUO
9892
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O60641
Snap91
Fusion protein corresponding to residues near the C terminal of human synaptosomal-associated protein, 91kDa.
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni