missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantità:
250 g
500 g
Descrizione
Sodium Molybdate Dihydrate, Extra Pure, SLR
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Colore | White |
pH | 9.0 to 10 |
Forma fisica | Powder/Solid |
Quantità | 250 g |
Peso formulazione | 241.95g/mol |
Imballaggio | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'