Learn More
Sodium Phosphate Tribasic Dodecahydrate, ≥98.0% (T), Honeywell Fluka™
Catalog Number 15686860
Click to view available options
Quantità:
250 g
1 kg
Imballaggio:
Poly bottle
10101-89-0
H24Na3O16P
380.119
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
10101-89-0
H24Na3O16P
380.119
MFCD00149198
ASTWEMOBIXQPPV-UHFFFAOYSA-K
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
61473
trisodium;phosphate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]
Puriss. p.a., ACS Reagent,≥98.0% (T)
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Sodium phosphate tribasic dodecahydrate
Punto di fusione ∼75°C
Colore White
Forma fisica Crystalline
Quantità 250 g
Formula lineare Na3PO4 · 12H2O
Numero UN NONH for all modes of transport
Informazioni di solubilità 70g/L at 20°C
Peso formulazione 380.12g/mol
Percent Purity ≥98.0% (T)
Imballaggio Poly bottle
Densità 1.62g/mL at 25°C; Bulk Density: 900 kg/m3