Learn More
Sofosbuvir impurity M, MedChemExpress
MedChemExpress Sofosbuvir impurity M, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Marca: Medchem Express HY-I0735
Descrizione
Sofosbuvir impurity M, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.Specifica
Sofosbuvir impurity M | |
Light Yellow | |
C22H30N3O10P | |
O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@@](O)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 | |
527.46 | |
Research | |
COVID-19-anti-virus |
2095551-10-1 | |
Research | |
5 mg | |
527.46 | |
99.04% | |
Solid |